• USA Home
  • 247391 - Methyl-[5-(N-methyl-N-phenylamino)-2,4-pentadienylidene]phenylammonium chloride

EMAIL THIS PAGE TO A FRIEND
247391 Sigma-Aldrich

Methyl-[5-(N-methyl-N-phenylamino)-2,4-pentadienylidene]phenylammonium chloride

Dye content 90 %

Purchase

Properties

composition   Dye content, 90%
mp   117-121 °C (lit.)
SMILES string   [Cl-].CN(\C=C\C=C\C=[N+](/C)c1ccccc1)c2ccccc2
InChI   1S/C19H21N2.ClH/c1-20(18-12-6-3-7-13-18)16-10-5-11-17-21(2)19-14-8-4-9-15-19;/h3-17H,1-2H3;1H/q+1;/p-1
InChI key   FPYATAHHYUQBFA-UHFFFAOYSA-M

Description

Application

Tricarbocyanine laser dye intermediate

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BZ1500000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?