• USA Home
  • 247413 - Bis(4-chlorophenyl) diselenide

EMAIL THIS PAGE TO A FRIEND
247413 Sigma-Aldrich

Bis(4-chlorophenyl) diselenide

98%

Synonym: 4,4′-Dichlorodiphenyl diselenide, 4-Chlorophenyl diselenide

  • CAS Number 20541-49-5

  • Linear Formula (ClC6H4)2Se2

  • Molecular Weight 381.02

  •  Beilstein/REAXYS Number 1974490

  •  EC Number 243-864-6

  •  MDL number MFCD00000599

Purchase

Properties

assay   98%
mp   85-87 °C (lit.)
SMILES string   Clc1ccc([Se][Se]c2ccc(Cl)cc2)cc1
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3283 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?