• USA Home
  • 248215 - (Dimethylamino)sulfur trifluoride

EMAIL THIS PAGE TO A FRIEND
248215 Aldrich

(Dimethylamino)sulfur trifluoride

≥95%

Synonym: Methyl DAST

  • CAS Number 3880-03-3

  • Linear Formula (CH3)2NSF3

  • Molecular Weight 133.14

  •  Beilstein Registry Number 1901089

  •  MDL number MFCD00009900

  •  PubChem Substance ID 24854921

Purchase

Properties

Related Categories Chemical Synthesis, Fluorination, Fluorination Reagents, Synthetic Reagents
InChI Key   SAVQQRYWWAGSQW-UHFFFAOYSA-N
assay   ≥95%
form   liquid
bp   117.5 °C(lit.)
mp   −78.7 °C(lit.)
density   1.364 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Packaging

5, 25 g in poly bottle

Application

Fluorinating Agent used in the fluorination of compounds such as
• Alchols, aldehydes and ketones
• Fluorinated Podophyllotoxin derivatives
• Protected glycosyl fluorides from thioglycosides

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGI
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?