• USA Home
  • 248258 - 5-Amino-2-chlorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
248258 Sigma-Aldrich

5-Amino-2-chlorobenzoic acid

85%, technical grade

  • CAS Number 89-54-3

  • Linear Formula H2NC6H3(Cl)CO2H

  • Molecular Weight 171.58

  •  Beilstein Registry Number 2090422

  •  EC Number 201-916-5

  •  MDL number MFCD00007845

  •  PubChem Substance ID 24854924

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
grade   technical grade
InChI Key   GVCFFVPEOLCYNN-UHFFFAOYSA-N
assay   85%
mp   184-188 °C (lit.)

Description

Packaging

100, 500 g in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?