• USA Home
  • 250325 - 3,6-Diphenyl-5-methylisoxazolo[4,5-c]pyridin-4(5H)-one

EMAIL THIS PAGE TO A FRIEND
250325

3,6-Diphenyl-5-methylisoxazolo[4,5-c]pyridin-4(5H)-one

97%

  • CAS Number 60986-80-3

  • Empirical Formula (Hill Notation) C19H14N2O2

  • Molecular Weight 302.33

  •  MDL number MFCD00010061

Purchase

Properties

assay   97%
SMILES string   CN1C(=O)c2c(C=C1c3ccccc3)onc2-c4ccccc4
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?