• USA Home
  • 250333 - 4-Hydroxy-3-(α-iminobenzyl)-1-methyl-6-phenylpyridin-2(1H)-one

EMAIL THIS PAGE TO A FRIEND
250333 Sigma-Aldrich

4-Hydroxy-3-(α-iminobenzyl)-1-methyl-6-phenylpyridin-2(1H)-one

99%

  • CAS Number 60986-58-5

  • Empirical Formula (Hill Notation) C19H16N2O2

  • Molecular Weight 304.34

  •  MDL number MFCD00010112

Purchase

Properties

assay   99%
mp   244-247 °C (lit.)
SMILES string   CN1C(=O)C(=C(O)C=C1c2ccccc2)C(=N)c3ccccc3
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?