• USA Home
  • 250503 - Dimethyl diethylmalonate

EMAIL THIS PAGE TO A FRIEND
250503 Sigma-Aldrich

Dimethyl diethylmalonate

99%

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   99%
refractive index   n20/D 1.427 (lit.)
bp   97 °C/17 mmHg (lit.)
density   1.04 g/mL at 25 °C (lit.)
storage temp.   room temp
SMILES string   CCC(CC)(C(=O)OC)C(=O)OC
InChI   1S/C9H16O4/c1-5-9(6-2,7(10)12-3)8(11)13-4/h5-6H2,1-4H3
InChI key   AYBLPISRXMEMBV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?