EMAIL THIS PAGE TO A FRIEND
250716 Sigma-Aldrich

Geranylacetone

Synonym: 6,10-Dimethyl-5,9-undecadien-2-one

  • CAS Number 3796-70-1

  • Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2COCH3

  • Molecular Weight 194.31

  •  Beilstein/REAXYS Number 1722277

  •  EC Number 223-269-8

  •  MDL number MFCD00008910

  •  PubChem Substance ID 24855069

Purchase

Properties

impurities   mixture of cis and trans isomer
refractive index   n20/D 1.467 (lit.)
bp   254-258 °C (lit.)
density   0.873 g/mL at 25 °C (lit.)
SMILES string   C\C(C)=C\CC\C(C)=C\CCC(C)=O
InChI   1S/C13H22O/c1-11(2)7-5-8-12(3)9-6-10-13(4)14/h7,9H,5-6,8,10H2,1-4H3/b12-9+
InChI key   HNZUNIKWNYHEJJ-FMIVXFBMSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?