• USA Home
  • 251518 - Tributyltin ethoxide

EMAIL THIS PAGE TO A FRIEND
251518 Sigma-Aldrich

Tributyltin ethoxide

97%

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
assay   97%
refractive index   n20/D 1.467 (lit.)
bp   92 °C/0.1 mmHg (lit.)
density   1.098 g/mL at 25 °C (lit.)
SMILES string   CCCC[Sn](CCCC)(CCCC)OCC
InChI   1S/3C4H9.C2H5O.Sn/c3*1-3-4-2;1-2-3;/h3*1,3-4H2,2H3;2H2,1H3;/q;;;-1;+1
InChI key   SDTAGBUVRXOKAK-UHFFFAOYSA-N

Description

Packaging

25, 100 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1992 6.1(3) / PGIII
WGK Germany 
3
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?