• USA Home
  • 251577 - 3-(Phenylsulfonyl)propionic acid

EMAIL THIS PAGE TO A FRIEND
251577 Sigma-Aldrich

3-(Phenylsulfonyl)propionic acid

99%

  • CAS Number 10154-71-9

  • Linear Formula C6H5SO2CH2CH2CO2H

  • Molecular Weight 214.24

  •  MDL number MFCD00010141

  •  PubChem Substance ID 24855141

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
assay   99%
mp   128-130 °C (lit.)
solubility   methanol: soluble 25 mg/mL, clear, colorless
SMILES string   OC(=O)CCS(=O)(=O)c1ccccc1
InChI   1S/C9H10O4S/c10-9(11)6-7-14(12,13)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)
InChI key   WGTYYNCSWCKXAI-UHFFFAOYSA-N

Description

Application

3-(Phenylsulfonyl)propionic acid may be used in chemical synthesis.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?