• USA Home
  • 252158 - N,5-Dimethyl-3-phenyl-4-isoxazolecarboxamide

EMAIL THIS PAGE TO A FRIEND
252158

N,5-Dimethyl-3-phenyl-4-isoxazolecarboxamide

99%

  • CAS Number 60986-88-1

  • Empirical Formula (Hill Notation) C12H12N2O2

  • Molecular Weight 216.24

  •  MDL number MFCD00010071

Purchase

Properties

assay   99%
SMILES string   CNC(=O)c1c(C)onc1-c2ccccc2
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?