• USA Home
  • 253227 - 4-(2-Bromoacetamido)-TEMPO

EMAIL THIS PAGE TO A FRIEND
253227 Sigma-Aldrich

4-(2-Bromoacetamido)-TEMPO

for ESR-spectroscopy

Synonym: 4-(2-Bromoacetamido)-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical

  • CAS Number 24567-97-3

  • Empirical Formula (Hill Notation) C11H20BrN2O2

  • Molecular Weight 292.19

  •  MDL number MFCD00010170

  •  PubChem Substance ID 24855303

Purchase

Properties

Related Categories Chemical Synthesis, Radical Chemistry, Stable Radicals, Synthetic Reagents, TEMPO Derivatives More...
grade   for ESR-spectroscopy
description   free radical
mp   118-120 °C (lit.)
SMILES string   CC1(C)CC(CC(C)(C)N1[O])NC(=O)CBr
InChI   1S/C11H20BrN2O2/c1-10(2)5-8(13-9(15)7-12)6-11(3,4)14(10)16/h8H,5-7H2,1-4H3,(H,13,15)
InChI key   RAOLMQDPNYDCEF-UHFFFAOYSA-N

Description

General description

Spin label.

Packaging

10 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?