• USA Home
  • 253863 - 1,3,5-Triethylbenzene

EMAIL THIS PAGE TO A FRIEND
253863 Sigma-Aldrich

1,3,5-Triethylbenzene

technical grade, 90%

  • CAS Number 102-25-0

  • Linear Formula C6H3(C2H5)3

  • Molecular Weight 162.27

  •  Beilstein Registry Number 2039481

  •  EC Number 203-017-3

  •  MDL number MFCD00009261

  •  PubChem Substance ID 24855335

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
grade   technical grade
InChI Key   WJYMPXJVHNDZHD-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.495(lit.)
bp   215 °C(lit.)
density   0.862 g/mL at 25 °C(lit.)

Description

Packaging

25 g in glass bottle

Application

1,3,5-Triethylbenzene was used in extraction of HReO4, HClO4 and HAuCl4 by trioctylphosphine oxide. It was also used in synthesis of 1,3,5-tris(pyrazol-1-ylmethyl)-2,4,6-triethylbenzene.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?