• USA Home
  • 256633 - B-Bromo-9-BBN solution

EMAIL THIS PAGE TO A FRIEND
256633 Sigma-Aldrich

B-Bromo-9-BBN solution

1.0 M in methylene chloride

Synonym: 9-Bromo-9-borabicyclo[3.3.1]nonane solution

  • CAS Number 22086-45-9

  • Empirical Formula (Hill Notation) C8H14BBr

  • Molecular Weight 200.91

  •  MDL number MFCD00074773

Purchase

Properties

concentration   1.0 M in methylene chloride
density   1.315 g/mL at 25 °C
SMILES string   BrB1C2CCCC1CCC2
InChI   1S/C8H14BBr/c10-9-7-3-1-4-8(9)6-2-5-7/h7-8H,1-6H2/t7-,8+
InChI key   FYZDPIIMSZAKGP-OCAPTIKFSA-N

Description

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

Safety & Documentation

Safety Information

RIDADR 
UN 2845 4.2/PG 1
Flash Point(F) 
1 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

septum inlet adapter

Product #

Description

Add to Cart

Z118206 Angled septum-inlet adapter, brass, inlet I.D. 6 mm × 6 mm
Z118141 Straight septum-inlet adapter, inlet I.D. 6 mm, brass
Z118192 Straight septum-inlet adapter, inlet I.D. 13.9 mm, brass

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?