• USA Home
  • 256994 - 4-Fluorophenylmagnesium bromide solution

EMAIL THIS PAGE TO A FRIEND
256994 Sigma-Aldrich

4-Fluorophenylmagnesium bromide solution

2.0 M in diethyl ether

  • CAS Number 352-13-6

  • Linear Formula FC6H4MgBr

  • Molecular Weight 199.30

  •  Beilstein/REAXYS Number 636802

  •  MDL number MFCD00009667

Purchase

Properties

concentration   2.0 M in diethyl ether
density   1.043 g/mL at 25 °C
SMILES string   Fc1ccc([Mg]Br)cc1
InChI   1S/C6H4F.BrH.Mg/c7-6-4-2-1-3-5-6;;/h2-5H;1H;/q;;+1/p-1
InChI key   QYBFFRXNNFXREA-UHFFFAOYSA-M

Description

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Reacts violently with water.
RIDADR 
UN 2924 8(3) / PGI
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

245550 4-Fluorophenylmagnesium bromide solution, 2.0 M in diethyl ether

septum inlet adapter

Product #

Description

Add to Cart

Z118206 Angled septum-inlet adapter, brass, inlet I.D. 6 mm × 6 mm
Z118141 Straight septum-inlet adapter, inlet I.D. 6 mm, brass

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?