• USA Home
  • 258318 - Hematin from porcine

EMAIL THIS PAGE TO A FRIEND
258318

Hematin from porcine

  • CAS Number 15489-90-4

  • Empirical Formula (Hill Notation) C34H33FeN4O5

  • Molecular Weight 633.49

Purchase
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?