• USA Home
  • 258636 - 4-Fluoro-3-nitrophenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
258636 Sigma-Aldrich

4-Fluoro-3-nitrophenyl isocyanate

98%

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C7, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   KCHOHFPMTUPOJU-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.5684 (lit.)
mp   27-28 °C (lit.)
density   1.455 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3335
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?