EMAIL THIS PAGE TO A FRIEND
258725 Sigma-Aldrich

Palmitic acid

99%

Synonym: 1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

  • CAS Number 57-10-3

  • Linear Formula CH3(CH2)14COOH

  • Molecular Weight 256.42

  •  Beilstein/REAXYS Number 607489

  •  EC Number 200-312-9

  •  MDL number MFCD00002747

Purchase

Properties

vapor pressure   10 mmHg ( 210 °C)
assay   99%
bp   271.5 °C/100 mmHg (lit.)
mp   61-62.5 °C (lit.)
density   0.852 g/mL at 25 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCC(O)=O
InChI   1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)
InChI key   IPCSVZSSVZVIGE-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RT4550000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P0500 Palmitic acid, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?