• USA Home
  • 259055 - 4-Methoxy-1-methylindole

EMAIL THIS PAGE TO A FRIEND
259055 Sigma-Aldrich

4-Methoxy-1-methylindole

98%

  • CAS Number 7556-35-6

  • Empirical Formula (Hill Notation) C10H11NO

  • Molecular Weight 161.20

  •  MDL number MFCD00010679

  •  PubChem Substance ID 24855682

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   ATEWBFOJLQMYEA-UHFFFAOYSA-N
assay   98%
mp   89-91 °C (lit.)

Description

Application

4-Methoxy-1-methylindole has been used:
• as internal standard for the quantification of indole-3-carbinol acetate, indole-3-carbinol and 3,3′-diindolylmethane in mouse plasma, liver and kidney tissues by HPLC
• in preparation of ethyl 9-methoxy-3,4,5-trimethylpyrrolo[3,2-b]carbazole-2-carboxylate

Reactant for:
• Preparation of GSK-3β inhibitors
• Palladium-catalyzed C2-arylation reactions

Packaging

500 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?