• USA Home
  • 25905 - (S)-(+)-2-Chloro-1-phenylethanol

EMAIL THIS PAGE TO A FRIEND
25905 Sigma-Aldrich

(S)-(+)-2-Chloro-1-phenylethanol

≥97.0% (sum of enantiomers, GC)

Synonym: (S)-(+)-α-(Chloromethyl)benzenemethanol

  • CAS Number 70111-05-6

  • Linear Formula C6H5CH(CH2Cl)OH

  • Molecular Weight 156.61

  •  Beilstein/REAXYS Number 3648988

  •  MDL number MFCD00077692

  •  PubChem Substance ID 57648154

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
assay   ≥97.0% (sum of enantiomers, GC)
optical activity   [α]20/D +48±2°, c = 2.8% in cyclohexane
optical purity   enantiomeric ratio: ≥98.5:1.5 (GC)
refractive index   n20/D 1.550 (lit.)
  n20/D 1.552
density   1.185 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   O[C@H](CCl)c1ccccc1
InChI   1S/C8H9ClO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6H2/t8-/m1/s1
InChI key   XWCQSILTDPAWDP-MRVPVSSYSA-N

Description

Packaging

1 mL in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?