• USA Home
  • 259136 - N,N′-Bis(2-aminoethyl)-1,3-propanediamine

EMAIL THIS PAGE TO A FRIEND
259136

N,N′-Bis(2-aminoethyl)-1,3-propanediamine

technical grade

  • CAS Number 4741-99-5

  • Linear Formula H2NCH2CH2NH(CH2)3NHCH2CH2NH2

  • Molecular Weight 160.26

  •  Beilstein/REAXYS Number 1742732

  •  MDL number MFCD00008174

Purchase

Properties

grade   technical grade
refractive index   n20/D 1.494 (lit.)
bp   142-145 °C/8 mmHg (lit.)
density   0.96 g/mL at 25 °C (lit.)
SMILES string   NCCNCCCNCCN
InChI   1S/C7H20N4/c8-2-6-10-4-1-5-11-7-3-9/h10-11H,1-9H2
InChI key   UWMHHZFHBCYGCV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?