• USA Home
  • 259438 - Phenyltrimethylammonium methosulfate

EMAIL THIS PAGE TO A FRIEND
259438 Aldrich

Phenyltrimethylammonium methosulfate

99%

  • CAS Number 28001-58-3

  • Empirical Formula (Hill Notation) C10H17NO4S

  • Molecular Weight 247.31

  •  MDL number MFCD00011792

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?