EMAIL THIS PAGE TO A FRIEND
259799 Sigma-Aldrich

Tetraethylsilane

97%

  • CAS Number 631-36-7

  • Linear Formula Si(C2H5)4

  • Molecular Weight 144.33

  •  Beilstein/REAXYS Number 1732768

  •  MDL number MFCD00009019

Purchase

Properties

assay   97%
refractive index   n20/D 1.4263 (lit.)
bp   153-154 °C (lit.)
mp   −82.5 °C (lit.)
density   0.761 g/mL at 25 °C (lit.)
SMILES string   CC[Si](CC)(CC)CC
InChI   1S/C8H20Si/c1-5-9(6-2,7-3)8-4/h5-8H2,1-4H3
InChI key   VCZQFJFZMMALHB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

510874 Tetraethylsilane, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?