EMAIL THIS PAGE TO A FRIEND
260142 Sigma-Aldrich

Apocholic acid

96%

Synonym: 3α,12α-Dihydroxy-5β,8(14)-cholen-24-oic acid, 3α,12α-Dihydroxy-5β-chol-8(14)en-24-oic acid, 5β,8(14)-Cholen-24-oic acid-3α,12α-diol

  • CAS Number 641-81-6

  • Empirical Formula (Hill Notation) C24H38O4

  • Molecular Weight 390.56

  •  MDL number MFCD00003622

  •  PubChem Substance ID 24855723

Purchase

Properties

assay   96%
optical activity   [α]25/D +45.0°, c = 1 in methanol
mp   175-176 °C (lit.)
SMILES string   C[C@H](CCC(O)=O)[C@H]1CCC2=C3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?