• USA Home
  • 260991 - Cobalt(III) sepulchrate trichloride

EMAIL THIS PAGE TO A FRIEND
260991 Sigma-Aldrich

Cobalt(III) sepulchrate trichloride

95%

Synonym: 1,3,6,8,10,13,16,19-Octaazabicyclo[6.6.6]eicosanecobalt trichloride

  • CAS Number 71963-57-0

  • Empirical Formula (Hill Notation) C12H30Cl3CoN8

  • Molecular Weight 451.71

  •  MDL number MFCD00013152

  •  PubChem Substance ID 24855783

Purchase

Properties

assay   95%
mp   262 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?