EMAIL THIS PAGE TO A FRIEND
261297 Aldrich

Ytterbium

powder (under oil), −40 mesh, 99.9%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
InChI Key   NAWDYIZEMPQZHO-UHFFFAOYSA-N
assay   99.9%
form   powder (under oil)
resistivity   28 μΩ-cm, 20°C
particle size   −40 mesh
bp   1196 °C(lit.)
mp   824 °C (lit.)
density   6.54 g/mL at 25 °C(lit.)

Description

Packaging

2 g in poly bottle

Application

When reacted with aromatic aldimines, followed by oxidants, produces vicinal diimine compounds.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3089 4.1 / PGII
WGK Germany 
3
RTECS 
ZG1925000
Protocols & Articles

Articles

Metal Hydrides for NiMH Battery Applications

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications. The Partnership for a New Generation of V...
Dhanesh Chandra,* Wen-Ming Chien and Anjali Talekar
Material Matters Volume 6 Article 2
Keywords: Absorption, Automotive, Capillary electrophoresis, Combustion, Degradations, Electrochemical analysis, Electronics, Environmental, Ion-exchange chromatography, Phase transitions, Precipitation, Purification, Separation, Substitutions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?