• USA Home
  • 261629 - 2-Naphthyl phosphate calcium salt hydrate

EMAIL THIS PAGE TO A FRIEND
261629 Sigma-Aldrich

2-Naphthyl phosphate calcium salt hydrate

98%

  • CAS Number 305808-24-6

  • Linear Formula [C10H7OP(O)(OH)O]2Ca · xH2O

  • Molecular Weight 262.21 (anhydrous basis)

  •  MDL number MFCD00149269

  •  PubChem Substance ID 24855825

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Phosphates/Phosphites, Phosphorus Compounds More...
assay   98%
mp   254-257 °C (lit.)
SMILES string   O.O=P1(O[Ca]O1)Oc2ccc3ccccc3c2
InChI   1S/2C10H9O4P.Ca.H2O/c2*11-15(12,13)14-10-6-5-8-3-1-2-4-9(8)7-10;;/h2*1-7H,(H2,11,12,13);;1H2/q;;+2;/p-2
InChI key   QGKCAHUPARGGEP-UHFFFAOYSA-L
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?