• USA Home
  • 261882 - 4-(2,4-Dichlorophenoxy)butyric acid

EMAIL THIS PAGE TO A FRIEND
261882 Sigma-Aldrich

4-(2,4-Dichlorophenoxy)butyric acid

95%

Synonym: 2,4-DB

  • CAS Number 94-82-6

  • Linear Formula Cl2C6H3O(CH2)3CO2H

  • Molecular Weight 249.09

  •  Beilstein/REAXYS Number 1976809

  •  EC Number 202-366-9

  •  MDL number MFCD00002819

  •  PubChem Substance ID 24855841

Purchase

Properties

InChI Key   YIVXMZJTEQBPQO-UHFFFAOYSA-N
assay   95%
mp   118-120 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
ES9100000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

45420 2,4-DB, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?