EMAIL THIS PAGE TO A FRIEND
262943 Sigma-Aldrich

Silicon monoxide

granular, −10-+10 mesh

Purchase

Properties

InChI Key   LIVNPJMFVYWSIS-UHFFFAOYSA-N
form   granular
particle size   −10-+10 mesh
density   2.13 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?