EMAIL THIS PAGE TO A FRIEND
264083 Sigma-Aldrich

Indium

bars, ≥99.999%

Synonym: Indium element

Purchase

Properties

vapor pressure   <0.01 mmHg ( 25 °C)
assay   ≥99.999%
form   bars
resistivity   8.37 μΩ-cm
mp   156.6 °C (lit.)
density   7.3 g/mL at 25 °C (lit.)
SMILES string   [In]
InChI   1S/In
InChI key   APFVFJFRJDLVQX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL1050000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

264091 Indium, rod, diam. 6 mm, ≥99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?