• USA Home
  • 264547 - 2-Methoxydibenzofuran

EMAIL THIS PAGE TO A FRIEND
264547 Aldrich

2-Methoxydibenzofuran

96%

  • CAS Number 20357-70-4

  • Empirical Formula (Hill Notation) C13H10O2

  • Molecular Weight 198.22

  •  MDL number MFCD00012313

Purchase

Properties

assay   96%
mp   42-45 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?