• USA Home
  • 264792 - 3-Mercapto-2-butanol, mixture of isomers

EMAIL THIS PAGE TO A FRIEND
264792 Sigma-Aldrich

3-Mercapto-2-butanol, mixture of isomers

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.4779 (lit.)
bp   59-61 °C/10 mmHg (lit.)
density   0.999 g/mL at 25 °C (lit.)
SMILES string   CC(O)C(C)S
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?