EMAIL THIS PAGE TO A FRIEND
265853 Sigma-Aldrich

Lead

rod, diam. 5.0 mm, 99.9995%

Synonym: Lead element

Purchase

Properties

assay   99.9995%
form   rod
resistivity   20.65 μΩ-cm
diam.   5.0 mm
bp   1740 °C (lit.)
mp   327.4 °C (lit.)
SMILES string   [Pb]
InChI   1S/Pb
InChI key   WABPQHHGFIMREM-UHFFFAOYSA-N

Description

Quantity

22 g = 100 mm

Safety & Documentation

Safety Information

WGK Germany 
-
RTECS 
OF7525000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?