EMAIL THIS PAGE TO A FRIEND
266337 Sigma-Aldrich

Antimony

lumps and powder, 99.8%

Synonym: Antimony black, Antimony element

Purchase

Properties

InChI Key   WATWJIUSRGPENY-UHFFFAOYSA-N
assay   99.8%
form   lumps and powder
resistance   41.7 μΩ-cm
bp   1635 °C (lit.)
mp   630 °C (lit.)
  630 °C (lit.)
density   6.69 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2871 6.1 / PGIII
WGK Germany 
2
RTECS 
CC4025000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

266329 Antimony, powder, -100 mesh, 99.5% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?