EMAIL THIS PAGE TO A FRIEND
266736 Aldrich

Copper

foil, thickness 0.5 mm, 99.999%

Purchase

Properties

InChI Key   RYGMFSIKBFXOCR-UHFFFAOYSA-N
assay   99.999%
form   foil
resistivity   1.673 μΩ-cm, 20°C
thickness   0.5 mm
bp   2567 °C(lit.)
mp   1083.4 °C(lit.)
density   8.94 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9 / PGIII
WGK Germany 
nwg
RTECS 
GL5325000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

266744 Copper, foil, thickness 1.0 mm, 99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?