EMAIL THIS PAGE TO A FRIEND
266884 Sigma-Aldrich

Iron

foil, thickness 1.0 mm, ≥99.99%

Purchase

Properties

InChI Key   XEEYBQQBJWHFJM-UHFFFAOYSA-N
assay   ≥99.99%
form   foil
resistivity   9.71 μΩ-cm
thickness   1.0 mm
bp   2750 °C (lit.)
mp   1535 °C (lit.)
density   7.86 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
nwg
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

338141 Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?