EMAIL THIS PAGE TO A FRIEND
267015 Sigma-Aldrich

Nickel

foil, thickness 1.0 mm, 99.995%

Purchase

Properties

assay   99.995%
form   foil
resistivity   6.97 μΩ-cm, 20°C
thickness   1.0 mm
bp   2732 °C (lit.)
mp   1453 °C (lit.)
density   8.9 g/mL at 25 °C (lit.)
SMILES string   [Ni]
InChI   1S/Ni
InChI key   PXHVJJICTQNCMI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
QR5950000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

357553 Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?