• USA Home
  • 268666 - 1-Aminoethylphosphonic acid

EMAIL THIS PAGE TO A FRIEND
268666 Sigma-Aldrich

1-Aminoethylphosphonic acid

96%

  • CAS Number 6323-97-3

  • Empirical Formula (Hill Notation) C2H8NO3P

  • Molecular Weight 125.06

  •  MDL number MFCD00008068

Purchase

Properties

assay   96%
SMILES string   CC(N)P(O)(O)=O
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?