• USA Home
  • 269069 - N-(4-Aminobutyl)-N-ethylisoluminol

EMAIL THIS PAGE TO A FRIEND
269069 Aldrich

N-(4-Aminobutyl)-N-ethylisoluminol

Synonym: 4-(N1-Ethyl-4-aminobutylamino)phthalic hydrazide, 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI

  • CAS Number 66612-29-1

  • Empirical Formula (Hill Notation) C14H20N4O2

  • Molecular Weight 276.33

  •  Beilstein/REAXYS Number 920895

  •  MDL number MFCD00010559

Purchase

Properties

InChI Key   LEOJISUPFSWNMA-UHFFFAOYSA-N
impurities   <15% water
mp   259-260 °C (lit.)

Description

Features and Benefits

Efficient chemiluminescent NH2-coupling reagent for detection of a wide variety of proteins down to the picomole range.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A0156 N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?