• USA Home
  • 269530 - Dimethyl (3-phenoxyacetonyl)phosphonate

EMAIL THIS PAGE TO A FRIEND
269530 Aldrich

Dimethyl (3-phenoxyacetonyl)phosphonate

97%

  • CAS Number 40665-68-7

  • Empirical Formula (Hill Notation) C11H15O5P

  • Molecular Weight 258.21

  •  MDL number MFCD00010225

Purchase

Properties

assay   97%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?