EMAIL THIS PAGE TO A FRIEND
270091 Sigma-Aldrich

NB-Enantride

Synonym: Lithium hydrido(9-BBN-nopol benzyl ether adduct), 0.5M in tetrahydrofuran solution

  • CAS Number 81572-37-4

  • Empirical Formula (Hill Notation) C26H40BLiO

  • Molecular Weight 386.35

  •  MDL number MFCD00135987

  •  PubChem Substance ID 24856353

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Reduction
density   0.947 g/mL at 25 °C (lit.)
SMILES string   [Li+].CC1(C)[C@@H]2C[C@H]([BH-]3[C@H]4CCC[C@@H]3CCC4)[C@@H](CCOCc5ccccc5)[C@H]1C2
InChI   1S/C26H40BO.Li/c1-26(2)20-16-24(26)23(14-15-28-18-19-8-4-3-5-9-19)25(17-20)27-21-10-6-11-22(27)13-7-12-21;/h3-5,8-9,20-25,27H,6-7,10-18H2,1-2H3;/q-1;+1/t20-,21-,22+,23-,24+,25-,27?;/m0./s1
InChI key   XJOCNAQVRHLSGK-QUERJQDASA-N

Description

Legal Information

NB-Enantride is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Reacts violently with water.
RIDADR 
UN 3399A 4.3(3) / PGI
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?