• USA Home
  • 270210 - Trimethylsilyl P,P-diethylphosphonoacetate

EMAIL THIS PAGE TO A FRIEND
270210 Sigma-Aldrich

Trimethylsilyl P,P-diethylphosphonoacetate

Synonym: Diethyl (trimethylsilyloxycarbonylmethyl)phosphonate, Phosphonoacetic acid P,P-diethyl trimethylsilyl ester

  • CAS Number 66130-90-3

  • Linear Formula (C2H5O)2P(O)CH2CO2Si(CH3)3

  • Molecular Weight 268.32

  •  Beilstein/REAXYS Number 2371028

  •  EC Number 266-171-0

  •  MDL number MFCD00009080

  •  PubChem Substance ID 24856362

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Horner-Wadsworth-Emmons Reagents, Olefination, Synthetic Reagents More...
refractive index   n20/D 1.430 (lit.)
bp   93 °C/0.05 mmHg
density   1.059 g/mL at 25 °C (lit.)
SMILES string   CCOP(=O)(CC(=O)O[Si](C)(C)C)OCC
InChI   1S/C9H21O5PSi/c1-6-12-15(11,13-7-2)8-9(10)14-16(3,4)5/h6-8H2,1-5H3
InChI key   VHIDZGBMGSRVHR-UHFFFAOYSA-N

Description

Application

Used in the synthesis of cytovaricin.

Packaging

5, 25 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
59 °F
Flash Point(C) 
15 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?