• USA Home
  • 270857 - Triethyl orthobenzoate

EMAIL THIS PAGE TO A FRIEND
270857 Sigma-Aldrich

Triethyl orthobenzoate

97%

  • CAS Number 1663-61-2

  • Linear Formula C6H5C(OC2H5)3

  • Molecular Weight 224.30

  •  Beilstein/REAXYS Number 645341

  •  EC Number 216-771-3

  •  MDL number MFCD00009222

Purchase

Properties

assay   97%
refractive index   n20/D 1.472 (lit.)
bp   239-241 °C (lit.)
  80 °C/0.1 mmHg (lit.)
density   0.991 g/mL at 25 °C (lit.)
SMILES string   CCOC(OCC)(OCC)c1ccccc1
InChI   1S/C13H20O3/c1-4-14-13(15-5-2,16-6-3)12-10-8-7-9-11-12/h7-11H,4-6H2,1-3H3
InChI key   BQFPCTXLBRVFJL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C
Protocols & Articles
Related Products

replacement part

Product #

Description

Add to Cart

75570 Triethyl orthobenzoate, ≥95.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?