• USA Home
  • 272701 - 5,6,7,8-Tetrahydroquinoxaline

EMAIL THIS PAGE TO A FRIEND
272701 Sigma-Aldrich

5,6,7,8-Tetrahydroquinoxaline

96%

Synonym: Cyclohexapyrazine, Tetrahydroquinoxaline

Purchase

Properties

assay   96%
refractive index   n20/D 1.542 (lit.)
bp   85 °C/3 mmHg (lit.)
density   1.061 g/mL at 25 °C (lit.)
SMILES string   C1CCc2nccnc2C1
InChI   1S/C8H10N2/c1-2-4-8-7(3-1)9-5-6-10-8/h5-6H,1-4H2
InChI key   XCZPDOCRSYZOBI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

W332100 5,6,7,8-Tetrahydroquinoxaline, ≥97%, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?