EMAIL THIS PAGE TO A FRIEND
272930 Sigma-Aldrich

4-(Benzyloxy)phenol

≥99%

Synonym: 4-(Phenylmethoxy)phenol, Hydroquinone monobenzyl ether

  • CAS Number 103-16-2

  • Linear Formula C6H5CH2OC6H4OH

  • Molecular Weight 200.23

  •  Beilstein/REAXYS Number 1958305

  •  EC Number 203-083-3

  •  MDL number MFCD00002333

  •  PubChem Substance ID 24856519

Purchase

Properties

assay   ≥99%
mp   119-120 °C (lit.)
SMILES string   Oc1ccc(OCc2ccccc2)cc1
InChI   1S/C13H12O2/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9,14H,10H2
InChI key   VYQNWZOUAUKGHI-UHFFFAOYSA-N
Gene Information   human ... TYR(7299)
rat ... Ar(24208)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

158348 4-(Benzyloxy)phenol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?