EMAIL THIS PAGE TO A FRIEND
274143 Sigma-Aldrich

DL-Lysine

98%

Synonym: (±)-2,6-Diaminocaproic acid

  • CAS Number 70-54-2

  • Linear Formula NH2(CH2)4CH(NH2)COOH

  • Molecular Weight 146.19

  •  Beilstein/REAXYS Number 1616991

  •  EC Number 200-740-6

  •  MDL number MFCD00064432

  •  PubChem Substance ID 329752993

Purchase

Properties

assay   98%
mp   170 °C (dec.) (lit.)
SMILES string   NCCCCC(N)C(O)=O
InChI   1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)
InChI key   KDXKERNSBIXSRK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
Protocols & Articles

Articles

HPLC Analysis of Lysine Enantiomers on Astec® CHIROBIOTIC® T (Ethanol Mobile Phase)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Lysine Enantiomers on Astec® CHIROBIOTIC® T (Ethanol Mobile Phase)
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?