• USA Home
  • 274224 - 4-Fluoro-4′-hydroxybenzophenone

EMAIL THIS PAGE TO A FRIEND
274224 Sigma-Aldrich

4-Fluoro-4′-hydroxybenzophenone

97%

Purchase

Description

Application

4-Fluoro-4′-hydroxybenzophenone was used to study the synthetic route of xerogels.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?