• USA Home
  • 274887 - (4-Cyanobenzyl)triphenylphosphonium chloride

EMAIL THIS PAGE TO A FRIEND
274887 Aldrich

(4-Cyanobenzyl)triphenylphosphonium chloride

98%

  • CAS Number 20430-33-5

  • Empirical Formula (Hill Notation) C26H21ClNP

  • Molecular Weight 413.88

  •  Beilstein/REAXYS Number 4630137

  •  MDL number MFCD00011752

Purchase

Properties

assay   98%
mp   ≥300 °C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?