EMAIL THIS PAGE TO A FRIEND
275514 Sigma-Aldrich

6-Aza-2-thiothymine

99%

Synonym: 2-Thio-6-azathymine, 3,4-Dihydro-6-methyl-3-thioxo-1,2,4-triazin-5(2H)-one

  • CAS Number 615-76-9

  • Empirical Formula (Hill Notation) C4H5N3OS

  • Molecular Weight 143.17

  •  Beilstein/REAXYS Number 4861

  •  EC Number 210-445-4

  •  MDL number MFCD00006458

  •  PubChem Substance ID 24856651

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Triazines
InChI Key   NKOPQOSBROLOFP-UHFFFAOYSA-N
assay   99%
mp   218-221 °C (lit.)
solubility   formic acid: soluble 5%, clear, colorless to light yellow-green

Description

Application

6-Aza-2-thiothymine was used in the preparation of a matrix that was utilized for ionizing small molecules.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XZ2853000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?