EMAIL THIS PAGE TO A FRIEND
27553 Sigma-Aldrich

(−)-Clovene

technical, ≥90% (sum of enantiomers, GC)

  • CAS Number 469-92-1

  • Empirical Formula (Hill Notation) C15H24

  • Molecular Weight 204.35

  •  Beilstein/REAXYS Number 2044272

  •  MDL number MFCD00210036

  •  PubChem Substance ID 57648258

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
Quality Level   100
grade   technical
assay   ≥90% (sum of enantiomers, GC)
optical activity   [α]20/D −27.5±1°, c = 1% in chloroform
refractive index   n20/D 1.495
storage temp.   2-8°C
SMILES string   C[C@@]12CCC[C@]3(C1)C=CC(C)(C)[C@H]3CC2
InChI   1S/C15H24/c1-13(2)9-10-15-7-4-6-14(3,11-15)8-5-12(13)15/h9-10,12H,4-8,11H2,1-3H3/t12-,14+,15-/m1/s1
InChI key   MKZIRHIVARSBHI-VHDGCEQUSA-N

Description

Packaging

100 mg in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?